Learn More
SNRPN upstream reading frame, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a highly basic protein localized to the nucleus. The evolutionarily constrained open reading frame is found on a bicistronic transcript which has a downstream ORF encoding the small nuclear ribonucleoprotein polypeptide N. The upstream coding region utilizes the first three exons of the transcript, a region that has been identified as an imprinting center. Multiple transcription initiation sites have been identified and extensive alternative splicing occurs in the 5' untranslated region but the full-length nature of these transcripts has not been determined. An alternate exon has been identified that substitutes for exon 4 and leads to a truncated, monocistronic transcript. Alternative splicing or deletion caused by a translocation event in the 5' UTR or coding region of this gene leads to Angelman syndrome or Prader-Willi syndrome due to parental imprint switch failure. The function of this protein is not yet known. [provided by RefSeq]
Specifications
Specifications
| Antigen | SNRPN upstream reading frame |
| Applications | Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human SNURF protein. |
| Formulation | No additive |
| Gene | SNURF |
| Gene Accession No. | BC024777 |
| Gene Symbols | SNURF |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.