Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX1 Mouse anti-Human, Unlabeled, Clone: 51, BD

Catalog No. BDB611482 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611482 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611482 Supplier BD Biosciences Supplier No. 611482
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Biological processes such as transmembrane signaling and receptor mediated endocytosis revolve around the function of cell surface receptors. A network of molecular machinery directs the intracellular trafficking of receptors during their biosynthesis and mediates signaling downstream of receptors. The sorting nexins (SNX1, SNX1A, SNX2, SNX3, and SNX4) are a family of intracellular proteins that are thought to direct the sorting of receptor proteins. The SNX proteins contain a conserved 100 amino acid region termed the phox homology (PX)
domain and are part of a family of hydrophilic proteins which includes S. cerevisiae proteins that function in protein sorting. SNX1, SNX2 and SNX4 associate predominantly with membranes and bind transmembrane receptors such as those for EGF, PDGF, and insulin. SNX1 directs the EGF receptor to the lysosomes for degradation. SNX2 forms homomeric complexes and heteromeric complexes with SNX1, SNX1A, and SNX4. These complexes are thought to be necessary for efficient protein sorting. Thus, SNX1 and other sorting nexins are thought to play important roles in the specificity of protein trafficking to and from the plasma membrane.

Host Species: Mouse
Clone: 51
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human SNX1 aa. 1-108
Formula Weight [Chemical]: 74kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX1
Applications Western Blot
Classification Monoclonal
Clone 51
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human SNX1 aa. 1-108
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.