Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sorting nexin 15 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89007511 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-511 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-511 Supplier Abnova Corporation Supplier No. H00029907B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SNX15 protein.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. This gene encodes two transcript variants encoding distinct isoforms. [provided by RefSeq

Sequence: MSRQAKDDFLRHYTVSDPRTHPKGYTEYKVTAQFISKKDPEDVKEVVVWKRYSDFRKLHGDLAYTHRNLFRRLEEFPAFPRAQVFGRFEASVIEERRKGAEDLLRFTVHIPALNNSPQLKEFFRGGEVTRPLEVSRDLHILPPPLIPTPPPDDPRLSQLLPAERRGLEELEVPVDPPPSSPAQEALDLLFNCESTEEASGSPARGPLTEAELALFDPFSKEEGAAPSPTHVAELATMEVESARLDQEPWEPGGQEEEEDGEGGPTPAYLSQATELITQALRDEKAGAYAAALQGYRDGVHVLLQGVPSDPLPARQEGVKKKAAEYLKRAEEILRLHLSQLPP

Specifications

Antigen sorting nexin 15
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SNX15 protein.
Formulation No additive
Gene SNX15
Gene Accession No. BC009897.1
Gene Alias HSAF001435
Gene Symbols SNX15
Host Species Mouse
Immunogen SNX15 (AAH09897.1, 1 a.a. ∼ 342 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 29907
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.