Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sorting nexin 4, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016953 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-953 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-953 Supplier Abnova Corporation Supplier No. H00008723B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SNX4 protein.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associated with the long isoform of the leptin receptor and with receptor tyrosine kinases for platelet-derived growth factor, insulin, and epidermal growth factor in cell cultures, but its function is unknown. This protein may form oligomeric complexes with family members. [provided by RefSeq

Sequence: MEQAPPDPERQLQPAPLEPLGSPDAGLGAAVGKEAEGAGEESSGVDTMTHNNFWLKKIEISVSEAEKRTGRNAMNMQETYTAYLIETRSVEHTDGQSVLTDSLWRRYSEFELLRSYLLVYYPHIVVPPLPEKRAEFVWHKLSADNMDPDFVERRRIGLENFLLRIASHPILCRDKIFYLFLTQEGNWKETVNETGFQLKADSRLKALNATFRVKNPDKRFTDLKHYSDELQSVISHLLRVRARVADRLYGVYKVHGNYGRVFSEWSAIEKEMGDGLQSAGHHMDVYASSIDDILEDEEHYADQLKEYLFYAEALRAVCRKHELMQYDLEMAAQDLASKKQQCEELVTGTVRTFSLKGMTTKLFGQETPEQREARIKVLEEQINEGEQQLKSKNLEGREFVKNAWADIERFKEQKNRDLKEALISYAVMQISMCKKGIQVWTNAKECFSKM

Specifications

Antigen sorting nexin 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SNX4 protein.
Formulation PBS with no preservative; pH 7.4
Gene SNX4
Gene Accession No. NM_003794
Gene Symbols SNX4
Host Species Mouse
Immunogen SNX4 (NP_003785.1, 1 a.a. ∼ 450 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8723
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.