Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS1 Mouse anti-Human, Clone: 4H1, MilliporeSigma™

Catalog No. 04002CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-002-CMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-002-CMI Supplier MilliporeSigma Supplier No. 04002C

Mouse Monoclonal Antibody

This Anti-SOCS1 Antibody, clone 4H1 (Ascites Free) is validated for use in western blotting and immunoprecipitation for the detection of SOCS1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4H1
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001258532
Gene Symbols Socs1, Cish1, Ssi1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to mouse SOCS1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.