Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS1 Mouse anti-Human, Mouse, Clone: 4H1, MilliporeSigma™

Catalog No. 04002MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-002-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-002-MI Supplier MilliporeSigma Supplier No. 04002
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SOCS1 Antibody, clone 4H1 is an antibody against SOCS1 for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4H1
Conjugate Unconjugated
Gene Accession No. NM_003745.1
Gene Symbols SOCS1, SSI-1, Cish1, TIP-3, TIP3, CISH1, SSI1, CIS1, SOCS-1, JAB
Host Species Mouse
Immunogen mouse SOCS1 recombinant GST fusion protein encompassing amino acids 2-78.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2 years at 2-8°C from date of shipment.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.