Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS3 Mouse anti-Human, Mouse, Clone: 1B2, MilliporeSigma™

Catalog No. 04004MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-004-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-004-MI Supplier MilliporeSigma Supplier No. 04004
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-SOCS3 Antibody, clone 1B2 is validated for use in western blotting for the detection of SOCS3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS3
Applications Western Blot
Classification Monoclonal
Clone 1B2
Conjugate Unconjugated
Formulation Protein G Purified mouse monoclonal IgG in a solution of 50% glycerol, 0.02MPhosphate buffer, pH 7.6, 0.25MNaCl, 0.1% sodium azide as a preservative.
Gene Accession No. NM_003955.3
Gene Symbols SOCS3, SSI3, ATOD4, MGC71791, SOCS-3, Cish3, SSI-3, CIS-3, CIS3
Host Species Mouse
Immunogen mouse SOCS3 recombinant GST fusion protein encompassing amino acids 2-44.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.