Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535075
Description
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| SOCS4 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q8WXH5 | |
| SOCS4 | |
| KLH conjugated synthetic peptide between 227-254 amino acids from the central region of human SOCS4 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| SOCS4 | |
| SOCS4; SOCS7; Suppressor of cytokine signaling 4; Suppressor of cytokine signaling 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 122809 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction