Learn More
Sodium-Calcium Exchanger 1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Sodium-Calcium Exchanger 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA. |
| Gene Symbols | SLC8A1; NCX1; DKFZp779F0871; +Ca; CNC; Na; MGC119581 |
| Host Species | Rabbit |
| Immunogen | A 23 amino acid peptide sequence within the cytoplasmic domain 3 rat NCX1 (Low et al. 1993; Furman et al. 1993; Lee et al. 1994; Nicoll et al. 1996; Kumoro et al. 1992; Blaustein & Lederer 1999). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.