Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Channel Nav1.5 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB549350UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB549350UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB549350UL Supplier MilliporeSigma Supplier No. AB549350UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Sodium Channel Nav1.5 Antibody, pain is an antibody against Sodium Channel Nav1.5 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sodium Channel Nav1.5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, and 0.05% sodium azide. Reconstitute with 50μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).
Gene Symbols SCN5A; HB2; ICCD; CMPD2; CMD1E; SSS1; HB1; PFHB1; IVF; HH1; Nav1.5; CDCD2; HBBD; LQT3
Host Species Rabbit
Immunogen Highly purified peptide corresponding to residues 493-511 of rat Nav1.5 (Accession P15389).
Purification Method Affinity Purified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000335.4; NM_001099404.1; NM_001099405.1; NM_198056.2
Target Species Rat
Content And Storage −20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.