Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Sodium channel Nav1.7 Mouse anti-Rat, Human, Clone: N68/6

Catalog No. MABN41MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN41MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN41MI Supplier MilliporeSigma™ Supplier No. MABN41
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Sodium channel Nav1.7 Antibody, clone N68/6 is validated for use in immunohistochemistry, western blotting for the detection of Sodium channel Nav1.7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sodium channel Nav1.7
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone N68/6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002968
Gene Symbols NENA, PN1, hNE-Na, FEB3B, NE-NA, ETHA, SCN9A
Host Species Mouse
Immunogen Recombinant protein corresponding to human Nav1.7.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.