Learn More
Description
Specifications
Specifications
| Antigen | Sodium Channel, Voltage Gated, Brain Type I |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody pre |
| Gene Symbols | SCN1A; SCN1; HBSCI; Nav1.1; FEB3; NAC1; SMEI; GEFSP2 |
| Host Species | Rabbit |
| Immunogen | Purified peptide from 465-481 of alpha subunit of rat type I voltage-gated sodium channel (VGSC) (Accession P04774). |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.