Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anti- Solanezumab (Anti-Idiotype) Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10124T10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10124T10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10124T10 Supplier R&D Systems Supplier No. FAB10124T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Solanezumab Monoclonal specifically detects Solanezumab in Multi-Species samples. It is validated for Flow Cytometry.

Specifications

Antigen Solanezumab
Applications Flow Cytometry
Classification Monoclonal
Clone 2372A
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Host Species Rabbit
Immunogen Full-length Solanezumab
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects Solanezumab in Western blots and direct ELISAs. In Western blots, no cross-reactivity with Trastuzumab and Adalimumab is observed.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.