Learn More
Sonic Hedgehog Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 061106
Description
Anti-Sonic Hedgehog Antibody detects level of Sonic Hedgehog and has been published and validated for use in Immunohistochemistry and Western Blotting.
Specifications
Sonic Hedgehog | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SHH; HHG-1; HHG1; HLP3; HPE3; MCOPCB5; SMMCI; TPT; TPTPS | |
GST-tagged recombinant protein corresponding to the N-terminus of Sonic HedgeHog. | |
100 μL | |
NP_000184 | |
2-8°C for 12 months |