Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Sortilin Clone: F11,

Catalog No. MABN1792MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1792MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1792MI Supplier MilliporeSigma™ Supplier No. MABN1792
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Sortilin Clone: F11 in Human, Mouse samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting

Sortilin (UniProt Q99523; also known as 100kDa NT receptor, Glycoprotein 95, Gp95, Neurotensin receptor 3, NT3, NTR3) is encoded by the SORT1 (also known as LDLCQ6, NT3) gene (Gene ID 6272) in human. Sortlin belongs to the family of sorting receptors that are characterized by the Vps10p domain, which creates a tunnel cavity with its ten-bladed β-propeller for the binding of soluble ligands. These family members also contain a short cytoplasmic tail that harbors recognition motifs for subcellular sorting adaptor proteins. Sortilin is reported to bind PCSK9 in the trans-Golgi network with high-affinity in HepG2 cells. Sort1 knockout mice exhibit low levels of PCSK9 in plasma and its subcellular localization in hepatocytes is also altered. Sortilin is also found to co-localize with amyloid precursor protein (APP) in neurites and it facilitates APP processing toward production of sAPPα, possibly by optimizing APP as substrate for α-secretase cleavage. Sortilin is initially produced with a signal (a.a. 1-33) and a propeptide (a.a. 34-77) sequence, the removal of which yields the mature receptor (a.a. 78-831) with a large luminal/extracellular (a.a. 78-555) portion, a transmembrane region(a.a. 756-778), and a short cytoplasmic tail (a.a.779-831). Like the other family members (MPRs and SorLA), sortilin resides both on the membrane of intracellular vesicles and on the plasma membrane. Altered sortilin expressions due to SORT1 genetic variations affect serum levels of low density lipoprotein cholesterol (LDL-C) and contribute to the LDL-C level quantitative trait locus 6 (LDLCQ6) and susceptibility to myocardial infarction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sortilin
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone F11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q99523
Gene Symbols SORT1
Host Species Mouse
Immunogen Recombinant protein corresponding to the extracellular domain of human Sortilin.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_002950.3
Target Species Human, Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.