Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sos1 Mouse, Unlabeled, Clone: 25, BD

Catalog No. BDB610096 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610096 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610096 Supplier BD Biosciences Supplier No. 610096

Mouse Monoclonal Antibody

The Sos (son of sevenless) gene was originally identified in Drosophila, and two mammalian homologues (mSos1 and mSos2) were isolated from a mouse cDNA library. These two cDNAs predict proteins that are approximately 70% identical in their amino acid residues. Both mSos1 and mSos2 are expressed in a wide number of mouse embryonic and adult tissues as well as in several cell lines. The human homologues, hSos1 and hSos2 have also been isolated and show a very high degree of amino acid identity to the mouse genes (98% for Sos1). Sos1 has a predicted molecular weight of 150kDa, but the apparent molecular weight is closer to 170kDa, presumably due to a high proline content. The mammalian Sos1 protein has a highly specific guanine nucleotide exchange activity toward p21ras . In EGF-stimulated cells, Sos1 interacts with the SH3 domains of GRB2, and GRB2 binds via its SH2 domain to tyrosine 1068 of the activated EGF receptor. Thus, GRB2 recruits Sos1 to the plasma membrane and enables it to activate the Ras signaling pathway by enhancing GTP loading on p21ras.

Host Species: Mouse
Clone: 25
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse mSos1 aa. 1-109
Formula Weight [Chemical]: 170kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sos1
Applications Western Blot
Classification Monoclonal
Clone 25
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse mSos1 aa. 1-109
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.