Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX-2 Mouse anti-Human, Mouse, Clone: 6F1.2, MilliporeSigma™

Catalog No. MAB4343MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4343MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4343MI Supplier MilliporeSigma Supplier No. MAB4343
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SOX-2 Antibody detects level of SOX-2 and has been published and validated for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX-2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6F1.2
Conjugate Unconjugated
Formulation Immunoglobulin purified from ascites by protein A affinity chromatography and presented as a liquid in 0.02Mphosphate buffer, pH 7.6, with 0.25MNaCl. Contains 0.1% sodium azide.
Gene Accession No. NM_003106.2
Gene Symbols SOX2, ANOP3, MGC2413, MCOPS3
Host Species Mouse
Immunogen Recombinant GST Fusion protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.