Learn More
SOX1 Mouse anti-Human, Unlabeled, Clone: N23-844, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 560749
Description
The N23-844 monoclonal antibody reacts with human SOX1, a member of the SOX (SRY-related HMG-box) family of transcription factors. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. It is one of the earliest transcription factors to be expressed in ectodermal cells committed to the neural fate. SOX1 is expressed in both embryonic and somatic neural stem and progenitor cells and it is down regulated during neuronal differentiation in many neuronal subtypes.
Bioimaging, Flow Cytometry, Western Blotting

Specifications
SOX1 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Peptide | |
0.1 mg | |
Cell Biology | |
Human | |
IgG1 κ |
Western Blot | |
N23-844 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.