Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox1 Mouse anti-Human, PerCP-Cy5.5, Clone: N23-844, BD

Catalog No. BDB561549 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561549 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561549 Supplier BD Biosciences Supplier No. 561549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The N23-844 monoclonal antibody reacts with human Sox1, a member of the SOX [SRY (sex determining region Y)-HMG-box] family of transcription factors. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. It is one of the earliest transcription factors to be expressed in ectodermal cells committed to the neural fate. Sox1 is expressed in both embryonic and somatic neural stem and progenitor cells, and it is down regulated during neuronal differentiation in many neuronal subtypes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox1
Applications Flow Cytometry
Classification Monoclonal
Clone N23-844
Conjugate PerCP-Cyanine5.5
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Sox1 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.