Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRY (sex determining region Y)-box 12, Mouse, Clone: 2A6, Abnova™

Catalog No. 89000972 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-972 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-972 Supplier Abnova Corporation Supplier No. H00006666M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SOX12.

Members of the SOX family of transcription factors are characterized by the presence of a DNA-binding high mobility group (HMG) domain, homologous to the HMG box of sex-determining region Y (SRY). Forming a subgroup of the HMG domain superfamily, SOX proteins have been implicated in cell fate decisions in a diverse range of developmental processes. SOX transcription factors have diverse tissue-specific expression patterns during early development and have been proposed to act as target-specific transcription factors and/or as chromatin structure regulatory elements. The protein encoded by this gene was identified as a SOX family member based on conserved domains and its expression in various tissues suggests a role in both differentiation and maintenance of several cell types. [provided by RefSeq

Sequence: LGFLSRLPPGPAGLDCSALDRDPDLQPPSGTSHFEFPDYCTPEVTEMIAGDWRPSSIADLVF

Specifications

Antigen SRY (sex determining region Y)-box 12
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SOX12.
Formulation PBS with no preservative; pH 7.4
Gene SOX12
Gene Accession No. NM_006943
Gene Alias SOX22
Gene Symbols SOX12
Host Species Mouse
Immunogen SOX12 (NP_008874, 252 a.a. ∼ 313 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6666
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.