Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox17 Mouse anti-Human, PE, Clone: P7-969, BD

Catalog No. BDB561591 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561591 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561591 Supplier BD Biosciences Supplier No. 561591
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The P7-969 monoclonal antibody reacts with human Sox17, a member of the SOX (SRY-releated HMG-box) family of transcription factors. SOX family members contain a DNA binding domain (HMG-box) and are involved in the control of development. Sox17 is expressed in primitive and definitive endoderm and regulates fetal and neonatal hematopoietic stem cell proliferation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox17
Applications Flow Cytometry
Classification Monoclonal
Clone P7-969
Conjugate PE
Description SOX-17, SOX17, FLJ22252
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Sox17 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.