Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB5603MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5603MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB5603MI Supplier MilliporeSigma Supplier No. AB5603
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Anti-SOX2 Antibody, Cat. No. AB5603, is a highly specific rabbit polyclonal antibody SOX2 and has been tested for use in Immunocytochemistry, and Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl, and 0.05% Sodium Azide, pH7.4.
Gene Symbols SOX2; ANOP3; MGC2413; MCOPS3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Sox2 RBX.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_003106.2
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.