Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox2 Mouse, Alexa Fluor 488, Clone: O30-678, BD

Catalog No. BDB561593 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561593 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561593 Supplier BD Biosciences Supplier No. 561593
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody O30-678 recognizes the Sox2 transcription factor. Sox2 [SRY (sex determining region Y)-box 2] is a member of the SRY-related HMG-box (SOX) family of transcription factors. Sox2 is required for the maintenance of the undifferentiated state of pluripotent stem cells. Complexes of Sox2 with the homeobox transcription factors Oct3/4 and/or Nanog bind to the promoters of a network of genes that are involved in the maintenance of pluripotency and self renewal in stem cells. Sox2 is also a marker of neural stem cells during embryonic development and in the adult brain. The O30-678 antibody recognizes both human and mouse Sox2 proteins.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox2
Applications Flow Cytometry
Classification Monoclonal
Clone O30-678
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Sox2 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.