Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox2 Mouse, Alexa Fluor 555, Clone: 245610, BD

Catalog No. BDB560293 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560293 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560293 Supplier BD Biosciences Supplier No. 560293

Mouse Monoclonal Antibody

The monoclonal antibody 245610 recognizes the Sox2 transcription factor. Sox2 (SRY (sex determining region Y)-box 2) is a member of the SRY-related HMG-box (SOX) family of transcription factors. Sox2 is required for the maintenance of the undifferentiated state of pluripotent stem cells. Complexes of Sox2 with the homeobox transcription factors Oct3/4 and/or Nanog bind to the promoters of a network of genes that are involved in the maintenance of pluripotency and self renewal in stem cells. The 245610 antibody recognizes both human and mouse proteins. Sox2 is also a marker of neural stem cells during embryonic development and in the adult brain.

Host Species: Mouse
Clone: 245610
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Human SOX2 Recombinant Protein

Bioimaging, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox2
Applications In vitro Assay
Classification Monoclonal
Clone 245610
Conjugate Alexa Fluor 555
Description ANOP3, MCOPS3, MGC2413
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human SOX2 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.