Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox3 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB5772MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5772MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5772MI Supplier MilliporeSigma Supplier No. AB5772

Rabbit Polyclonal Antibody

This Anti-Sox3 Antibody is validated for use in western blot, immunohistochemistry for the detection of Sox3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.
Gene Symbols SOX3; SOXB; MRGH
Host Species Rabbit
Immunogen Synthetic peptide, amino acids 283-295 from human Sox-3.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005634.2
Target Species Human, Mouse
Content And Storage -70°C for up to 6 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.