Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRY (sex determining region Y)-box 30, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018062 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-062 Supplier Abnova Corporation Supplier No. H00011063D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SOX30 protein.

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may be involved in the differentiation of developing male germ cells. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: MERARPEPPPQPRPLRPAPPPLPVEGTSFWAAAMEPPPSSPTLSAAASATLASSCGEAVASGLQPAVRRLLQVKPEQVLLLPQPQAQNEEAAASSAQARLLQFRPDLRLLQPPTASDGATSRPELHPVQPLALHVKAKKQKLGPSLDQSVGPRGAVETGPRASRVVKLEGPGPALGYFRGDEKGKLEAEEVMRDSMQGGAGKSPAAIREGVIKTEEPERLLEDCRLGAEPASNGLVHGSAEVILAPTSGAFGPHQQDLRIPLTLHTVPPGARIQFQGAPPSELIRLTKVPLTPVPTKMQSLLEPSVKIETKDVPLTVLPSDAGIPDTPFSKDRNGHVKRPMNAFMVWARIHRPALAKANPAANNAEISVQLGLEWNKLSEEQKKPYYDEAQKIKEKHREEFPGWVYQPRPGKRKRFPLSVSNVFSGTTQNIISTNPTTVYPYRSPTYSVVIPSLQNPITHPVVHALTVGLPLAMEIFRVQCQNALVIMKTGTQNMRVSFQL

Specifications

Antigen SRY (sex determining region Y)-box 30
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SOX30 protein.
Formulation PBS with no preservative; pH 7.4
Gene SOX30
Gene Accession No. NM_007017.2
Gene Symbols SOX30
Host Species Rabbit
Immunogen SOX30 (NP_008948.1, 1 a.a. ∼ 501 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 11063
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.