Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sox6 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5805
Description
Detect Sox6 with Anti-Sox6 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in western blot, immunocytochemistry.
Specifications
Sox6 | |
Polyclonal | |
Affinity purified immunoglobulin. Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunocytochemistry, Western Blot | |
Unconjugated | |
SOX6; HSSOX6; Sox-6 | |
Synthetic peptide, amino acids 349-364 of Sox6. | |
100 μg | |
NM_017508.1; NM_033326.2 | |
-70°C for up to 6 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction