Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox9 Rabbit anti-Human, Mouse, Rat, Chicken, Polyclonal, MilliporeSigma™

Catalog No. AB5535MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5535MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5535MI Supplier MilliporeSigma Supplier No. AB5535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Sox9 Antibody is a well characterized affinity purified Rabbit Polyclonal Antibody that reliably detects Transcription Factor Sox-9. This highly published antibody has been validated in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris -Glycine, 0.15M NaCl, 0.05% sodium azide, pH7.4.
Gene Symbols SOX9; CMPD1; CMD1; SRA1
Host Species Rabbit
Immunogen Synthetic peptide from human Sox9.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_000337
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.