Learn More
SRY (sex determining region Y)-box 9, Mouse, Clone: 3C10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SOX9.
Supplier: Abnova Corporation H00006662M02
Description
The protein encoded by this gene recognizes the sequence CCTTGAG along with other members of the HMG-box class DNA-binding proteins. It acts during chondrocyte differentiation and, with steroidogenic factor 1, regulates transcription of the anti-Muellerian hormone (AMH) gene. Deficiencies lead to the skeletal malformation syndrome campomelic dysplasia, frequently with sex reversal. [provided by RefSeq
Sequence: EQLSPSHYSEQQQHSPQQIAYSPFNLPHYSPSYPPITRSQYDYTDHQNSSSYYSHAAGQGTGLYSTFTYMNPAQRPMYTPIADTSGVPSIPQTHSPQHWEQPVYTQLTRPSpecifications
SRY (sex determining region Y)-box 9 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000346 | |
SOX9 | |
SOX9 (NP_000337, 400 a.a. ∼ 509 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
3C10 | |
Mouse monoclonal antibody raised against a partial recombinant SOX9. | |
SOX9 | |
CMD1/CMPD1/SRA1 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
6662 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.