Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sp1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07645MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-645-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-645-MI Supplier MilliporeSigma Supplier No. 07645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Sp1 Antibody (Rabbit Polyclonal Antibody) validated in Chimmunoprecipitation, EMSA, immunofluoresence, immunoprecipitation, western blotting to detect Sp1 also known as Sp1 transcription factor, specificity protein 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sp1
Applications ChIP Assay, Electromobility Shift Assay, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide and 30% glycerol.
Gene Symbols TSFP1
Host Species Rabbit
Immunogen Full length human Sp1 protein.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003100.1
Target Species Human, Mouse, Rat
Content And Storage −20°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.