Learn More
SP1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA532599
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
SP1 is a sequence-specific transcription factor that recognizes GGGGCGGGGC and closely related sequences, which are often referred to as GC boxes. SP1 belongs to a subgroup of transcription factors that are phosphorylated upon binding to promoter sequences.
Specifications
SP1 | |
Polyclonal | |
SP1 | |
specificity protein 1, transcription factor Sp1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P08047 | |
SP1 | |
Synthetic peptide derived from C-terminus of human SP1 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.