Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sperm associated antigen 6, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018174 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-174 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-174 Supplier Abnova Corporation Supplier No. H00009576D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SPAG6 protein.

The correlation of anti-sperm antibodies with cases of unexplained infertility implicates a role for these antibodies in blocking fertilization. Improved diagnosis and treatment of immunologic infertility, as well as identification of proteins for targeted contraception, are dependent on the identification and characterization of relevant sperm antigens. The protein expressed by this gene is recognized by anti-sperm antibodies from an infertile man. This protein localizes to the tail of permeabilized human sperm and contains eight contiguous armadillo repeats, a motif known to mediate protein-protein interactions. Studies in mice suggest that this protein is involved in sperm flagellar motility and maintenance of the structural integrity of mature sperm. Alternatively spliced variants that encode different protein isoforms have been described but the full-length sequences of only two have been determined. [provided by RefSeq

Sequence: MSQRQVLQVFEQYQKARTQFVQMVAELATRPQNIETLQNAGVMSLLRTLLLDVVPTIQQTAALALGRLANYNDDLAEAVVKCDILPQLVYSLAEQNRFYKKAAAFVLRAVGKHSPQLAQAIVDCGALDTLVICLEDFDPGVKEAAAWALRYIARHNAELSQAVVDAGAVPLLVLCIQEPEIALKRIAASALSDIAKHSPELAQTVVDAGAVAHLAQMILNPDAKLKHQILSALSQVSKHSVDLAEMVVEAEIFPVVLTCLKDKDEYVKKNASTLIREIAKHTPELSQLVVNAGGVAAVIDCIGSCKGNTRLPGIMMLGYVAAHSENLAMAVIISKGVPQLSVCLSEEPEDHIKAAAAWALGQIGRHTPEHARAVAVTNTLPVLLSLYMSTESSEDLQVKSKKAIKNILQKCTYLPALEPFLYDAPPNILKHVVGQFSKVLPHDSKARRLFVTSGGLKKVQEIKAEPGSLLQEYINSINSCYPEEIVRYYSPGYSDTLLQRVDSYQPLNN

Specifications

Antigen sperm associated antigen 6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SPAG6 protein.
Formulation PBS with no preservative; pH 7.4
Gene SPAG6
Gene Accession No. NM_012443.2
Gene Alias DKFZp434I153/MGC26276/Repro-SA-1/pf16
Gene Symbols SPAG6
Host Species Rabbit
Immunogen SPAG6 (NP_036575.1, 1 a.a. ∼ 509 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9576
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.