Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SPARC, Alexa Fluor 488, Clone: 124413, R&D Systems™

Catalog No. IC942G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC942G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC942G Supplier R&D Systems Supplier No. IC942G
This item is not returnable. View return policy

Rat Monoclonal Antibody

Applications:

Flow Cytometry (Intracellular Staining)

Specifications

Antigen SPARC
Applications Flow Cytometry
Classification Monoclonal
Clone 124413
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P07214
Gene Alias Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin)
Gene Symbols SPARC
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SPARC/Osteonectin Ala18-Ile302 Accession # P07214
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6678
Test Specificity Detects mouse SPARC/Osteonectin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human SPARC is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.