Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin alpha chain (nonerythroid) Mouse anti-Chicken, Guinea Pig, Human, Mouse, Rat, Rabbit, Pig, Mammals, Clone: AA6, MilliporeSigma™

Catalog No. MAB1622MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1622MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1622MI Supplier MilliporeSigma Supplier No. MAB1622
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Spectrin alpha chain (nonerythroid) Antibody, clone AA6 is an antibody against Spectrin alpha chain (nonerythroid) for use in immunohistochemistry, immunoprecipitation, western blot, Chimmunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spectrin alpha chain (nonerythroid)
Applications ChIP Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AA6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing PBS with 1% BSA and 0.09% sodium azide.
Gene Accession No. NM_003127.2
Gene Symbols SPTAN1, FLJ44613, SPTA2
Host Species Mouse
Immunogen Chicken red blood cell membranes purified by hypotonic lysis and mechanical enucleation.
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Chicken, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.