Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEG complex locus, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018215 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-215 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-215 Supplier Abnova Corporation Supplier No. H00010290D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SPEG protein.

Expression of this gene is thought to serve as a marker for differentiated vascular smooth muscle cells which may have a role in regulating growth and differentiation of this cell type. The encoded protein is highly similar to the corresponding rat and mouse proteins. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of only one variant has been defined. [provided by RefSeq

Sequence: MKPSPSQNRRSSDTGSKAPPTFKVSLMDQSVREGQDVIMSIRVQGEPKPVVSWLRNRQPVRPDQRRFAEEAEGGLCRLRILAAERGDAGFYTCKAVNEYGARQCEARLEVRGE

Specifications

Antigen SPEG complex locus
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SPEG protein.
Formulation PBS with no preservative; pH 7.4
Gene SPEG
Gene Accession No. BC006346.2
Gene Alias APEG1/BPEG/KIAA1297/MGC12676/SPEGalpha/SPEGbeta
Gene Symbols SPEG
Host Species Rabbit
Immunogen SPEG (AAH06346.1, 1 a.a. ∼ 113 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 10290
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.