Learn More
spastic paraplegia 21 (autosomal recessive, Mast syndrome), Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene was identified by a two-hybrid screen using CD4 as the bait. It binds to the hydrophobic C-terminal amino acids of CD4 which are involved in repression of T cell activation. The interaction with CD4 is mediated by the noncatalytic alpha/beta hydrolase fold domain of this protein. It is thus proposed that this gene product modulates the stimulatory activity of CD4. At least three different transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | spastic paraplegia 21 (autosomal recessive, Mast syndrome) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant SPG21. |
| Formulation | 50% glycerol |
| Gene | SPG21 |
| Gene Accession No. | NM_016630 |
| Gene Alias | ACP33/BM-019/GL010/MASPARDIN/MAST |
| Gene Symbols | SPG21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.