Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPG7, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002959 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-959 Supplier Abnova Corporation Supplier No. H00006687D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SPG7 protein.

This gene encodes a nuclear-encoded mitochondrial metalloprotease protein that is a member of the AAA (ATPases associated with a variety of cellular activities) protein family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. Two transcript variants encoding distinct isoforms have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 7. [provided by RefSeq

Sequence: MAVLLLLLRALRRGPGPGPRPLWGPGPAWSPGFPARPGRGRPYMASRPPGDLAEAGGRALQSLQLRLLTPTFEGINGLLLKQHLVQNPVRLWQLLGGTFYFNTSRLKQKNKEKDKSKGKAPEEDEEERRRRERDDQMYRERLRTLLVIAVVMSLLNALSTSGGSISWNDFVHEMLAKGEVQRVQVVPESDVVEVYLHPGAVVFGRPRLALMYRMQVANIDKFEEKLRAAEDELNIEAKDRIPVSYKRTGFFGNALYSVGMTAVGLAILWYVFRLAGMTGREGGFSAFNQLKMARFTIVDGKMGKGVSFKDVAGMHEAKLEVREFVDYLKSPERFLQLGAKVPKGALLLGPPGCGKTLLAKAVATEAQVPFLAMAGPEFVEVIGGLGAARVRSLFKEARARAPCIVYIDEIDAVGKKRSTTMSGFSNTEEEQTLNQLLVEMDGMGTTDHVIVLASTNRADILDGALMRPGRLDRHVFIDLPTLQERREIFEQHLKSLKLTQSSTFYSQRLAELTPGFSGADIANICNEAALHAAREGHTSVHTLNFEYAVERVLAGTAKKSKILSKEEQKVVAFHESGHALVGWMLEHTEAVMKVSITPRTNAALGFAQMLPRDQHLFTKEQLFERMCMALGGRASEALSFNEVTSGAQDDLRKVTRIAYSMVKQFGMAPGIGPISFPEAQEGLMGIGRRPFSQGLQQMMDHEARLLVAKAYRHTEKVLQDNLDKLQALANALLEKEVINYEDIEALIGPPPHGPKKMIAPQRWIDAQREKQDLGEEETEETQQPPLGGEEPTWPK

Specifications

Antigen SPG7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SPG7 protein.
Formulation PBS with no preservative; pH 7.4
Gene SPG7
Gene Accession No. NM_003119.2
Gene Alias CAR/CMAR/FLJ37308/MGC126331/MGC126332/PGN/SPG5C
Gene Symbols SPG7
Host Species Rabbit
Immunogen SPG7 (NP_003110.1, 1 a.a. ∼ 795 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6687
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.