Learn More
SGMS1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA112762
Description
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.
A suggested positive control for this product is brain, heart, kidney, liver, muscle or stomach lysate.
Specifications
SGMS1 | |
Polyclonal | |
Unconjugated | |
Q86VZ5 | |
SGMS1 | |
Full-length recombinant sphingomyelin synthase 1 fusion protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
1mg/mL | |
SGMS1 | |
MGC17342, MOB, MOB1, SMS1, TMEM23, hmob33 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
259230 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.