Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin), Mouse, Clone: 4G3, Abnova™

Catalog No. 89001547 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-547 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-547 Supplier Abnova Corporation Supplier No. H00057119M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SPINLW1.

This gene encodes an epididymal protease inhibitor, which contains both kunitz-type and WAP-type four-disulfide core (WFDC) protease inhibitor consensus sequences. Most WFDC genes are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene is a member of the WFDC gene family and belongs to the telomeric cluster. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq

Sequence: PGLTDWLFPRRCPKIREECEFQERDVCTKDRQCQDNKKCCVFSCGKKCLDLKQDVCEMPKETGPCLAYFLHWWYDKKDNTCSMFVYGGCQGNNNNFQSKANCLNTCKNKRFP

Specifications

Antigen serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SPINLW1.
Formulation ascites with no preservative
Gene SPINLW1
Gene Accession No. NM_020398
Gene Alias EPPIN/EPPIN1/EPPIN2/EPPIN3/WAP7/WFDC7/dJ461P17.2
Gene Symbols SPINLW1
Host Species Mouse
Immunogen SPINLW1 (NP_065131, 22 a.a. ∼ 133 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 57119
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.