Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spinophilin/Neurabin-II Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. 06852MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-852-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-852-MI Supplier MilliporeSigma Supplier No. 06852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Spinophilin/Neurabin-II Antibody is an antibody against Spinophilin/Neurabin-II for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spinophilin/Neurabin-II
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 70% storage buffer (0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide) and 30% glycerol. Store at −20°C.
Gene Symbols PPP1R9B; PPP1R6; SPINO; Spinophilin; PPP1R9; Neurabin-2; FLJ30345; spinophilin; Neurabin-II
Host Species Rabbit
Immunogen GST fusion protein corresponding to residues 286-390 of rat brain spinophilin/neurabin-II.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_032595
Target Species Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.