Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spinophilin Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5669MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5669MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5669MI Supplier MilliporeSigma Supplier No. AB5669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Spinophilin Antibody detects level of Spinophilin and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spinophilin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in 0.02M phosphate buffer containing 0.25M NaCl, pH 7.6 with 0.1% sodium azide.
Gene Symbols PPP1R9B; Spinophilin; PPP1R9; Neurabin-2; PPP1R6; FLJ30345; spinophilin; Neurabin-II; SPINO
Host Species Rabbit
Immunogen Synthetic peptide from rat Spinophilin.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_032595.3
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.