Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SR protein family Mouse anti-Human, Mouse, Rat, Clone: 16H3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE126
Description
Anti-SR protein family Antibody, clone 16H3 is a Mouse Monoclonal Antibody for detection of SR protein family also known as arginine/serine-rich splicing factor 6 and has been validated in western blot, immunoprecipitation, immunocytochemistry.
Specifications
SR protein family | |
Monoclonal | |
Unconjugated | |
NP_006266 | |
Mouse | |
Protein G Purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
16H3 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
SRP55, B52, SFRS6 | |
Dephosphorylated bovine SRp55 and GST-tagged recombinant dephosphorylated SRp55 (Neugebauer, KM et. al 1995). | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction