Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERAC1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524288
Description
This antibody is predicted to react with bovine based on sequence homology.
Specifications
| SERAC1 | |
| Polyclonal | |
| SERAC1 | |
| protein SERAC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 321007, 84947 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q96JX3, Q3U213 | |
| SERAC1 | |
| KLH conjugated synthetic peptide between 552-581 amino acids from the C-terminal region of human SRAC1 | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction