Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRC1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514515
Description
This antibody is predicted to react with porcine based on sequence homology.
SRC1 acts as a transcriptional coactivator for steroid and nuclear hormone receptors. It is a member of the p160/steroid receptor coactivator (SRC) family and like other family members has histone acetyltransferase activity and contains a nuclear localization signal, as well as bHLH and PAS domains. This protein binds nuclear receptors directly and stimulates the transcriptional activities in a hormone-dependent fashion.
Specifications
SRC1 | |
Polyclonal | |
NCOA1 | |
Nuclear receptor coactivator 1; NCoA-1; Steroid receptor coactivator 1; SRC-1; RIP160; Protein Hin-2; Renal carcinoma antigen NY-REN-52; NCOA1; SRC1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
8648 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q15788 | |
NCOA1 | |
KLH conjugated synthetic peptide between 186-214 amino acids from the N-terminal region of human SRC1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction