Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2), Mouse, Clone: 1F4, Abnova™

Catalog No. 89000978 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-978 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-978 Supplier Abnova Corporation Supplier No. H00006716M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SRD5A2.

This gene encodes a microsomal protein expressed at high levels in androgen-sensitive tissues such as the prostate. The encoded protein is active at acidic pH and is sensitive to the 4-azasteroid inhibitor finasteride. Deficiencies in this gene can result in male pseudohermaphroditism, specifically pseudovaginal perineoscrotal hypospadias (PPSH). [provided by RefSeq

Sequence: AKPSGYGKHTESLKPAATRLPARAAWFLQELPSFAVPA

Specifications

Antigen steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SRD5A2.
Formulation PBS with no preservative; pH 7.4
Gene SRD5A2
Gene Accession No. NM_000348
Gene Alias MGC138457
Gene Symbols SRD5A2
Host Species Mouse
Immunogen SRD5A2 (NP_000339.2, 28 a.a. ∼ 65 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6716
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.