Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRD5A3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525200
Description
SRD5A3 converts testosterone (T) into 5-alpha-dihydrotestosterone (DHT).
Specifications
SRD5A3 | |
Polyclonal | |
SRD5A3 | |
CDG1P, CDG1Q, KRIZI, SRD5A2L, SRD5A2L1, 3-oxo-5-alpha-steroid 4-dehydrogenase 3, S5AR 3, SR type 3, probable polyprenol reductase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79644 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9H8P0 | |
SRD5A3 | |
KLH conjugated synthetic peptide between 283-311 amino acids from the C-terminal region of human SRD5A3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction