Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sterol regulatory element binding transcription factor 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004322 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-322 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-322 Supplier Abnova Corporation Supplier No. H00006721A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SREBF2.

This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain. [provided by RefSeq

Sequence: QAFCKNLLERAIESLVKPQAKKKAGDQEEESCEFSSALEYLKLLHSFVDSVGVMSPPLSRSSVLKSALGPDIICRWWTSAITVAISWLQGDDAAVRSHFT

Specifications

Antigen sterol regulatory element binding transcription factor 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SREBF2.
Formulation 50% glycerol
Gene SREBF2
Gene Accession No. BC056158
Gene Alias SREBP2/bHLHd2
Gene Symbols SREBF2
Host Species Mouse
Immunogen SREBF2 (AAH56158, 801 a.a. ∼ 900 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6721
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.