Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Mouse anti-Human, Clone: 2121, MilliporeSigma™

Catalog No. 04469MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-469-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-469-MI Supplier MilliporeSigma Supplier No. 04469
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SREBP1 Antibody, clone 2121 is a Mouse Monoclonal Antibody for detection of SREBP1 also known as sterol regulatory element binding transcription factor 1 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SREBP1
Applications Western Blot
Classification Monoclonal
Clone 2121
Conjugate Unconjugated
Formulation 100μL (1mg/mL) of protein A purified mouse monoclonal IgG1 in PBS with 0.1% sodium azide.
Gene Accession No. NM_001005291.1; NM_004176.3
Gene Symbols SREBF1, SREBP1, SREBP-1
Host Species Mouse
Immunogen Recombinant His-tagged human SREBP-1a protein encompassing amino acids 301-407.
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.