Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRp30a (SF2) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 061363MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-136-3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-136-3MI Supplier MilliporeSigma Supplier No. 061363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-SRp30a (SF2) Antibody (rabbit polyclonal antibody) validated in western blot, immunocytochemistry to detect SRp30a (SF2) also known as serine/arginine-rich splicing factor 1, pre-mRNA-splicing factor SF2- P33 subunit, SR splicing factor 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRp30a (SF2)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols ASF-1; SF2; ASF; SFRS1; SF2p33; SRp30a; SRSF1; SF2P333
Host Species Rabbit
Immunogen Linear peptide corresponding to human SRp30a.
Purification Method Protein A
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001071634
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.