Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRp40 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 061365MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-136-5MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-136-5MI Supplier MilliporeSigma Supplier No. 061365
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-SRp40 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunoprecipitation to detect SRp40 also known as serine/arginine-rich splicing factor 5, Pre-mRNA-splicing factor SRP40.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRp40
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols SRP40; SRSF5; HRS; SFRS5
Host Species Rabbit
Immunogen Linear peptide corresponding to human SRp40.
Purification Method Protein A
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001034554
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.